Läs mer
(R)-Sulindac Methyl Ester, TRC
Produktkod. 30421580
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30421580 100 mg
1 options
Produktkod. 30421580 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS699305100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Sulindac Methyl Ester
CAS 929081-61-8
Molekylformel C21H19FO3S
Kvantitet 100 mg
Synonym (1Z)-5-Fluoro-2-methyl-1-[[4-[(R)-methylsulfinyl]phenyl]methylene]-1H-indene-3-acetic Acid Methyl Ester
InChI formel InChI=1S/C21H19FO3S/c1-13-18(10-14-4-7-16(8-5-14)26(3)24)17-9-6-15(22)11-20(17)19(13)12-21(23)25-2/h4-11H,12H2,1-3H3/b18-10-/t26-/m1/s1
LEDER COC(=O)CC1=C(C)\C(=C\c2ccc(cc2)[S@](=O)C)\c3ccc(F)cc13
IUPAC-namn methyl 2-[(3Z)-6-fluoro-2-methyl-3-[[4-[(R)-methylsulfinyl]phenyl]methylidene]inden-1-yl]acetate
Molekylvikt (g/mol) 370.44
Formel vikt 370.1039