Läs mer
R-Thioctic Acid Tromethamine Impurity, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Thioctic Acid Ethyl Ester |
| CAS | 46353-61-1 |
| Molekylformel | C10 H18 O2 S2 |
| Kvantitet | 250 mg |
| Synonym | R-Thioctic Acid Tromethamine Impurity, 1,2-Dithiolane-3-pentanoic acid ethyl ester, 1,2-Dithiolane-3-valeric acid','ethyl ester (7CI), Ethyl thioctate, Ethyl-DL-α-lipoate, α-Lipoic acid ethyl ester |
| InChI formel | InChI=1S/C10H18O2S2/c1-2-12-10(11)6-4-3-5-9-7-8-13-14-9/h9H,2-8H2,1H3 |
| LEDER | CCOC(=O)CCCCC1CCSS1 |
| IUPAC-namn | ethyl 5-(dithiolan-3-yl)pentanoate |
| Molekylvikt (g/mol) | 234.379 |
| Formel vikt | 234.075 |