Läs mer
(R)-(+)-Verapamil Hydrochloride, TRC
Produktkod. 30452602
Change view
Kvantitet:
2 mg
Produktkod. Kvantitet
30452602 2 mg
1 options
Produktkod. 30452602 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV1249902MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-(+)-Verapamil Hydrochloride
CAS 38176-02-2
Molekylformel C27 H38 N2 O4 . Cl H
Kvantitet 2 mg
Synonym (αR)-α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-(1-methylethyl)benzeneacetonitrile hydrochloride (1:1), (R)-α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-(1-methylethyl)benzeneacetonitrile monohydrochloride, (+)-Verapamil hydrochloride, (R)-Verapamil hydrochloride, Dexverapamil monohydrochloride, LU 33925, NSC 632821
InChI formel InChI=1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H/t27-;/m1./s1
LEDER Cl.COc1ccc(CCN(C)CCC[C@@](C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC
IUPAC-namn (2R)-2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
Molekylvikt (g/mol) 491.06
Formel vikt 490.2598