Läs mer
R-(+)-Warfarin, TRC
Produktkod. 30454724
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454724 10 mg
30451094 5 mg
30450581 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454724 Leverantör Toronto Research Chemicals Leverantörsnummer TRCW49849010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5543-58-8
Molekylformel C19 H16 O4
Molekylvikt (g/mol) 308.33
Synonym 2H-1-Benzopyran-2-one, 4-hydroxy-3-[(1R)-3-oxo-1-phenylbutyl]-, 2H-1-Benzopyran-2-one, 4-hydroxy-3-(3-oxo-1-phenylbutyl)-, (R)-, Coumarin, 3-(α-acetonylbenzyl)-4-hydroxy-, R-(+)- (8CI), 4-Hydroxy-3-[(1R)-3-oxo-1-phenylbutyl]-2H-1-benzopyran-2-one, (+)-(R)-Warfarin, (+)-Warfarin, (R)-Warfarin, Dextrowarfarin, R-(+)-Warfarin, Warfarin, (+)-
IUPAC-namn 4-hydroxy-3-[(1R)-3-oxo-1-phenylbutyl]chromen-2-one
LEDER CC(=O)C[C@@H](C1=C(O)c2ccccc2OC1=O)c3ccccc3
Specifikationer
Kemiskt namn eller material (R)-Warfarin
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3/t15-/m1/s1
Formel vikt 308.1049
Procent renhet >95%