Learn More
Reagecon™ R1 Funhsin Solution Decolorized
Brand: Reagecon™ 1039402
Additional Details : CAS Number : 569-61-9 Weight : 0.10000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Specifications
100 mL | |
Decolorized | |
Funhsin Solution Decolorized |
Safety and Handling
- FUCHSIN SOLUTION, DECOLOURISED R1
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid 37%, 4, 4'-(4-iminocyclohexa-2, 5-dienylidenemethylene)dianiline hydrochloride