Läs mer
R406, TRC
Produktkod. 30010156
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010156 10 mg
30006805 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010156 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR06000010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 841290-81-1
Molekylformel C22 H23 F N6 O5 . (C6 H6 O3 S)
Molekylvikt (g/mol) 628.6
Synonym 6-[[5-Fluoro-2-[(3,4,5-trimethoxyphenyl)amino]-4-pyrimidinyl]amino]-2,2-dimethyl-2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one Monobenzenesulfonate, R406
IUPAC-namn benzenesulfonic acid;6-[[5-fluoro-2-(3,4,5-trimethoxyanilino)pyrimidin-4-yl]amino]-2,2-dimethyl-4H-pyrido[3,2-b][1,4]oxazin-3-one
LEDER COc1cc(Nc2ncc(F)c(N=C3NC4=C(OC(C)(C)C(=O)N4)C=C3)n2)cc(OC)c1OC.OS(=O)(=O)c5ccccc5
Specifikationer
Kemiskt namn eller material Tamatinib Besilate
Kvantitet 10 mg
InChI formel InChI=1S/C22H23FN6O5.C6H6O3S/c1-22(2)20(30)28-19-13(34-22)6-7-16(27-19)26-18-12(23)10-24-21(29-18)25-11-8-14(31-3)17(33-5)15(9-11)32-4;7-10(8,9)6-4-2-1-3-5-6/h6-10H,1-5H3,(H3,24,25,26,27,28,29,30);1-5H,(H,7,8,9)
Formel vikt 628.1752