Läs mer
Tocris Bioscience™ RA 839
Nrf2 activator; inhibits Nrf2/Keap1 interaction
Brand: Tocris Bioscience™ 5707/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
RA 839 is a Nrf2 activator; inhibits Nrf2/Keap1 interaction (Kd = 6 μM). Suppresses LPS-induced iNOS and nitric oxide expression in macrophages. Anti-inflammatory.Specifikationer
| (3S)-1-[4-[[(2,3,5,6-Tetramethylphenyl)sulfonyl]amino]-1-naphthalenyl]-3-pyrrolidinecarboxylic acid | |
| 1832713-02-6 | |
| 50 mg |