Learn More
Epredia™ RA Lamb 14-Drawer Slide Filing Cabinets, mini, grey
Product Code. 12151830 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Gray
Slide Capacity:
6000
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Slide Capacity Unit Size
12151830 Gray 6000 Each
12131820 Blue 6000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12151830 Supplier Epredia™ Supplier No. E43/12GREY
  • ONLINE EXCLUSIVE PRICE
    3850.00 SEK
    3526.63 SEK / Paket med 1
    Save 323.37 SEK
    8% Av
View all promotions
I lager
Vertically store up to 6000 76 x 26mm microscope slides with these 14-Drawer Cabinets.
  • Capacity of 3000 or 6000 slides depending on model
TRUSTED_SUSTAINABILITY
Specifications
Type Slide Filing Cabinet
Slide Capacity 6000
Color Gray
Depth (Metric) Unit 480 mm
Device Holds 6000 Slides
Height (Metric) Unit 480 mm
Length (Metric) Slide 76 mm
Material Steel
Width (Metric) Slide 26 mm
Width (Metric) Unit 140 mm