missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RAB10 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825385
|
Proteintech
27094-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815385
|
Proteintech
27094-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.Specifications
RAB10 | |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P61026 | |
10890 | |
RAB10 Fusion Protein Ag25830 | |
Primary | |
-20°C | |
RAB10 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RAB10, Ras related protein Rab 10 | |
RAB10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |