Learn More
Abnova™ RAB2 Recombinant Protein
Brand:  Abnova™ H00005862-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16179961
  • 6845.00 SEK / 25µg
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH08929
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
49.06
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAYAYLFKYIIIGDTGVGKSCLLLQFTDKRFQPVHDLTIGVEFGARMITIDGKQIKLQIWDTAGQESFRSITRSYYRGAAGALLVYDITRRDTFNHLTTWLEDARQHSNSNMVIMLIGNKSDLESRREVKKEEGEAFAREHGLIFMETSAKTASNVEEAFINTAKEIYEKIQEGVFDINNEANGIKIGPQHAATNATHAGNQGGQQAGGGCC
RAB2
RAB2A
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
5862
RAB2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RAB2A
Human
Recombinant
Solution