missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3GAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24599-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the RAB3 protein family, members of which are involved in regulated exocytosis of neurotransmitters and hormones. This protein forms the Rab3 GTPase-activating complex with RAB3GAP1, where it constitutes the regulatory subunit, whereas the latter functions as the catalytic subunit. This gene has the highest level of expression in the brain, consistent with it having a key role in neurodevelopment. Mutations in this gene are associated with Martsolf syndrome.Specifications
RAB3GAP2 | |
Polyclonal | |
Unconjugated | |
RAB3GAP2 | |
KIAA0839, p150, Rab3 GAP p150, Rab3 GAP regulatory subunit, RAB3 GAP150, RAB3GAP150, RAB3GAP2, RGAP iso | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25782, 289350, 98732 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5U1Z0, Q8BMG7, Q9H2M9 | |
Rab3gap2 | |
RAB3GAP2 Fusion Protein Ag20254 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |