missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
RAB41 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18818-1-AP-150UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Rab 41 (Ras-related protein Rab-41) is a 222 amino acid protein that belongs to the small GTPase superfamily and the Rab family.Members of the Rab familyfunction asregulators ofmembrane trafficking. Theycycle between inactive GDP-bound and activated GTP-bound states,which iscontrolled by GTP hydrolysis-activating proteins(GAPs). Rab 41 can be activated bythe GAP protein RN-Tre.Localizing to the Golgicomplex, Rab 41 isinvolved in protein transport and participatesin GTP binding. Existing astwo alternativelyspliced isoforms, the gene that encodesthe Rab 41 proteinmapsto human chromosome Xq13.1. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes. Color blindness, hemophilia, and Duchennemuscular dystrophy arewellknown X chromosome-linked conditionswhich affectmales more frequently asmalescarry a single X chromosome.Specifikationer
| RAB41 | |
| Polyclonal | |
| Unconjugated | |
| RAB41 | |
| RAB41, Ras related protein Rab 41 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 347517 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5JT25 | |
| RAB41 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering