missing translation for 'onlineSavingsMsg'
Learn More

RAB9A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11420-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888483

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ras-related protein Rab-9A, also called RAB9, is a protein that in humans is encoded by the RAB9A gene. This gene is mapped to Xp22.2. RAB9 has been localized to components of the endocytic/exocytic pathway and has been implicated in recycling of membrane receptors. It has been found that downregulation of RAB9A gene expression in HeLa cells induced severe cell vacuolation. RAB9A GTPase is directly bound by TIP47 in its active, GTP-bound conformation. Moreover, RAB9A increases the affinity of TIP47 for its cargo. What′s more, this gene may involved in the transport of proteins between the endosomes and the trans Golgi network. RAB9A has been shown to interact with RABEPK and TIP47.
Specifications

Specifications

RAB9A
Polyclonal
Unconjugated
RAB9A
RAB9, RAB9A, Ras related protein Rab 9A
Rabbit
Antigen Affinity Chromatography
RUO
9367
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51151
RAB9A
RAB9A Fusion Protein Ag1969
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers