missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB9A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11420-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Ras-related protein Rab-9A, also called RAB9, is a protein that in humans is encoded by the RAB9A gene. This gene is mapped to Xp22.2. RAB9 has been localized to components of the endocytic/exocytic pathway and has been implicated in recycling of membrane receptors. It has been found that downregulation of RAB9A gene expression in HeLa cells induced severe cell vacuolation. RAB9A GTPase is directly bound by TIP47 in its active, GTP-bound conformation. Moreover, RAB9A increases the affinity of TIP47 for its cargo. What′s more, this gene may involved in the transport of proteins between the endosomes and the trans Golgi network. RAB9A has been shown to interact with RABEPK and TIP47.Specifications
RAB9A | |
Polyclonal | |
Unconjugated | |
RAB9A | |
RAB9, RAB9A, Ras related protein Rab 9A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9367 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51151 | |
RAB9A | |
RAB9A Fusion Protein Ag1969 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |