Läs mer
APOA5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Beskrivning
The protein encoded by this gene is an apolipoprotein and an important determinant of plasma triglyceride levels, a major risk factor for coronary artery disease. It is a component of high density lipoprotein and is highly similar to a rat protein that is upregulated in response to liver injury. This gene uses alternate polyadenylation sites and is located proximal to the apolipoprotein gene cluster on chromosome 11q23. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | APOA5 |
| Användningsområden | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of APOA5. |
| Utspädning | Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gen | APOA5 |
| Genaccessionsnr. | NP_443200.2;Q6Q788 |
| Gene Alias | APOA-V/APOAV/MGC126836/MGC126838/RAP3 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.