missing translation for 'onlineSavingsMsg'
Läs mer

AR (phospho S650), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16035267
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16035267 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16035267 Leverantör Abnova Leverantörsnummer PAB25030.100uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of AR.

The androgen receptor gene is more than 90kb long and codes for a protein that has 3 major functional domains: the N-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormone activated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This gene contains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in the N-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscular atrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS). Two alternatively spliced variants encoding distinct isoforms have been described. [provided by RefSeq

Specifikationer

Antigen AR
Användningsområden Immunohistochemistry (PFA fixed)
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of AR.
Utspädning Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulering In PBS, pH 7.2 (0.05% sodium azide)
Gen AR
Gene Alias AIS/DHTR/HUMARA/KD/NR3C4/SBMA/SMAX1/TFM
Gensymboler AR
Värd art Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S650 of AR.
Reningsmetod Antigen affinity purification
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 367
Målarter Human
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.