missing translation for 'onlineSavingsMsg'
Läs mer

BAD (phospho S134), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16058786
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
400 μL
Förpackningsstorlek:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16058786 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16058786 Leverantör Abnova Leverantörsnummer PAB0416.400uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of BAD.

The protein encoded by this gene is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform. [provided by RefSeq

Specifikationer

Antigen BAD
Användningsområden ELISA, Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of BAD.
Utspädning Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.09% sodium azide)
Gen BAD
Genaccessionsnr. NP_116784;Q92934
Gene Alias BBC2/BCL2L8
Gensymboler BAD
Värd art Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S134 of human BAD.
Reningsmetod Protein G purification
Kvantitet 400 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 572
Målarter Human
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.