missing translation for 'onlineSavingsMsg'
Läs mer

CBX1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16091117
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μg
Förpackningsstorlek:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16091117 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16091117 Leverantör Abnova Leverantörsnummer PAB14123.50ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against full-length recombinant CBX1.

This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family . The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Specifikationer

Antigen CBX1
Användningsområden Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against full length recombinant CBX1.
Utspädning Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.05% sodium azide, 0.05% proclin 300)
Gen CBX1
Genaccessionsnr. P83916
Gene Alias CBX/HP1-BETA/HP1Hs-beta/HP1Hsbeta/M31/MOD1/p25beta
Gensymboler CBX1
Värd art Rabbit
Immunogen Recombinant GST fusion protein corresponding to full length human CBX1.
Kvantitet 50 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 10951
Testspecificitet This antibody also recognizes CBX5 and CBX3.
Målarter Human
Innehåll och lagring Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.