missing translation for 'onlineSavingsMsg'
Läs mer

CDCA4, Rabbit, Polyclonal Antibody, Abnova™

Produktkod. 16092937
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16092937 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16092937 Leverantör Abnova Leverantörsnummer PAB17501.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of CDCA4.

This gene encodes a protein that belongs to the E2F family of transcription factors. This protein regulates E2F-dependent transcriptional activation and cell proliferation, mainly through the E2F/retinoblastoma protein pathway. It also functions in the regulaton of JUN oncogene expression. This protein shows distinctive nuclear-mitotic apparatus distribution, it is involved in spindle organization from prometaphase, and may also play a role as a midzone factor involved in chromosome segregation or cytokinesis. Two alternatively spliced transcript variants encoding the same protein have been noted for this gene. Two pseudogenes have also been identified on chromosome 1. [provided by RefSeq

Specifikationer

Antigen CDCA4
Användningsområden ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of CDCA4.
Utspädning Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulering In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gen CDCA4
Genaccessionsnr. Q9BXL8
Gene Alias FLJ20764/FLJ52878/HEPP/MGC19517/SEI-3/HEPP
Gensymboler CDCA4
Värd art Rabbit
Immunogen A synthetic peptide corresponding to internal of human CDCA4.
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 55038
Testspecificitet This antibody detects endogenous levels of total CDCA4 protein.
Målarter Human, Mouse
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.