Läs mer
CDK6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CDK6.
Brand: Abnova PAB28592.100uL
Beskrivning
The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This kinase is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression and G1/S transition. The activity of this kinase first appears in mid-G1 phase, which is controlled by the regulatory subunits including D-type cyclins and members of INK4 family of CDK inhibitors. This kinase, as well as CDK4, has been shown to phosphorylate, and thus regulate the activity of, tumor suppressor protein Rb. [provided by RefSeq
Sequence: FRGSSDVDQLGKILDVIGLPGEEDWPRDVALPRQAFHSKSAQPIEKFVTDIDELGKDLLLKCLTFNPAKRISAYSALSHPYFQDLERCKENLDSHLPPSQNTSELNSpecifikationer
| CDK6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CDK6. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| Q00534 | |
| CDK6 | |
| Recombinant protein corresponding to amino acids of human CDK6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/ml) Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| CDK6 | |
| MGC59692/PLSTIRE/STQTL11 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1021 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only