missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody, FITC

Produktkod. 11575180 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 μg
Förpackningsstorlek:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11575180 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11575180 Leverantör Invitrogen™ Leverantörsnummer SA19511

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Secondary Antibody

This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9511 immunogen is immunoglobulin isolated from chicken serum. Store at 2-8°C as an undiluted liquid for several weeks. After dilution, do not use for more than one day.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Chicken IgY (H+L)
Användningsområden Immunohistochemistry, Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera FITC
Formulering PBS with 25% glycerol, 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Värd art Rabbit
Immunogen Immunoglobulin isolated from chicken serum.
Reningsmetod Affinity chromatography
Kvantitet 500 μg
Regulatorisk status RUO
Primär eller sekundär Secondary
Målarter Chicken
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles, store in dark
Produkttyp Secondary Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.