Läs mer
CYSLTR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Beskrivning
The cysteinyl leukotrienes LTC4, LTD4, and LTE4 are important mediators of human bronchial asthma. Pharmacologic studies have determined that cysteinyl leukotrienes activate at least 2 receptors, the protein encoded by this gene and CYSLTR2. This encoded receptor is a member of the superfamily of G protein-coupled receptors. Activation of this receptor by LTD4 results in contraction and proliferation of smooth muscle, oedema, eosinophil migration and damage to the mucus layer in the lung. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | CYSLTR1 |
| Användningsområden | Immunofluorescence, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of CYSLTR1. |
| Utspädning | Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.2 (0.05% sodium azide) |
| Gen | CYSLTR1 |
| Gene Alias | CYSLT1/CYSLT1R/CYSLTR/HG55/HMTMF81/MGC46139 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.