missing translation for 'onlineSavingsMsg'
Läs mer

DOK1 (phospho Y362), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Produktkod. 16050738
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16050738 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16050738 Leverantör Abnova Leverantörsnummer PAB7920.100uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of DOK1.

Docking protein 1 is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia (CML) patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. Docking protein 1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs. Docking protein 2 binds p120 (RasGAP) from CML cells. It has been postulated to play a role in mitogenic signaling. [provided by RefSeq

Specifikationer

Antigen DOK1
Användningsområden ELISA, Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of DOK1.
Utspädning ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulering In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gen DOK1
Gene Alias MGC117395/MGC138860/P62DOK
Gensymboler DOK1
Värd art Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y362 of human DOK1.
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 1796
Testspecificitet This sequence is conserved in Dok1 from rat and mouse (Tyr-361), and has high homology to Dok2 (Tyr-337). The site is not conserved in other Dok family members.
Målarter Human, Mouse, Rat
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.