Läs mer
EGFR (phospho Y1092), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Beskrivning
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | EGFR |
| Användningsområden | Immunohistochemistry (PFA fixed), Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR. |
| Utspädning | Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gen | EGFR |
| Genaccessionsnr. | P00533 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.