Läs mer
EXO1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Beskrivning
This gene encodes a protein with 5' to 3' exonuclease activity as well as an RNase H activity. It is similar to the Saccharomyces cerevisiae protein Exo1 which interacts with Msh2 and which is involved in mismatch repair and recombination. Alternative splicing of this gene results in three transcript variants encoding two different isoforms. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | EXO1 |
| Användningsområden | Immunofluorescence, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of EXO1. |
| Utspädning | Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.2 (0.09% sodium azide) |
| Gen | EXO1 |
| Genaccessionsnr. | Q9UQ84 |
| Gene Alias | HEX1/hExoI |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.