missing translation for 'onlineSavingsMsg'
Läs mer

FRMPD2, Rabbit, Polyclonal Antibody, Abnova™

Produktkod. 16092437
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16092437 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16092437 Leverantör Abnova Leverantörsnummer PAB16852.100ug

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of FRMPD2.

This gene encodes a FERM and PDZ domain-containing protein and is located in a region of chromosome 10q which contains a segmental duplication resulting in three nearly identical regions. This copy of the gene is full-length and is in the telomeric duplicated region. The exact function of this gene is unknown; however, FERM domains often play a role in signal transduction pathways. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifikationer

Antigen FRMPD2
Användningsområden ELISA, Immunohistochemistry, Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of FRMPD2.
Utspädning Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.02% sodium azide)
Gen FRMPD2
Genaccessionsnr. NP_001018081
Gene Alias MGC35285/MGC87776/MGC87777/MGC90186/PDZD5C/PDZK5C
Gensymboler FRMPD2
Värd art Rabbit
Immunogen A synthetic peptide corresponding to N-terminus 15 amino acids of human FRMPD2.
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 143162
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.