missing translation for 'onlineSavingsMsg'
Läs mer

GHRHR Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16066407
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μg
Förpackningsstorlek:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16066407 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16066407 Leverantör Abnova Leverantörsnummer PAB26225.50ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of GHRHR.

This gene, expressed in the pituitary, encodes a receptor for growth hormone-releasing hormone. Binding of this hormone to the receptor leads to synthesis and release of growth hormone. Mutations in this gene have been associated with isolated growth hormone deficiency (IGHD), also known as Dwarfism of Sindh, a disorder characterized by short stature. Many alternate transcriptional splice variants encoding different isoforms have been described, but only two have been characterized to date. [provided by RefSeq

Specifikationer

Antigen GHRHR
Användningsområden Immunohistochemistry (PFA fixed)
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of GHRHR.
Utspädning Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.09% sodium azide)
Gen GHRHR
Genaccessionsnr. Q02643
Gene Alias GHRFR/GHRHRpsv/GRFR
Gensymboler GHRHR
Värd art Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acid at N-terminus of human GHRHR.
Reningsmetod Immunoaffinity chromatography
Kvantitet 50 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2692
Testspecificitet BLAST analysis of the peptide immunogen showed no homology with other human proteins, except FLJ23588 (50%).
Målarter Human
Innehåll och lagring Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.