Läs mer
GPR132 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Beskrivning
This gene encodes a subfamily member of the G-protein couple receptor (GPCR) superfamily. The encoded protein is a high-affinity receptor for lysophosphatidylcholine (LPC), a major phospholipid component of oxidized low density lipoprotein. This protein may react to LPC levels at sites of inflammation to limit the expansion of tissue-infiltrating cells. A similar protein in mouse is involved in cell cycle progression. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | GPR132 |
| Användningsområden | Immunohistochemistry (PFA fixed) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of GPR132. |
| Utspädning | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gen | GPR132 |
| Genaccessionsnr. | Q9UNW8 |
| Gene Alias | G2A/MGC99642 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.