missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
GPR153 Rabbit anti-Human, Monkey, Mouse, Polyclonal Antibody, Abnova™
Specifikationer
Specifikationer
| Antigen | GPR153 |
| Användningsområden | Immunohistochemistry (PFA fixed) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of GPR153. |
| Utspädning | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (34 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gen | GPR153 |
| Genaccessionsnr. | Q6NV75 |
| Gene Alias | DKFZp762B2210/PGR1 |
| Visa mer |
For Research Use Only
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?