Learn More
HDAC1 (phospho S421/S423), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Product Code. 16062517
Change view
Click to view available options
Kvantitet:
100 μg
Unit Size:
100µg
Product Code. Kvantitet unitSize
16062517 100 μg 100µg
1 options
Product Code. 16062517 Supplier Abnova Supplier No. PAB16973.100ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Rabbit polyclonal antibody raised against synthetic phosphopeptide of HDAC1.
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. The protein encoded by this gene belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, it deacetylates p53 and modulates its effect on cell growth and apoptosis. [provided by RefSeq
Specifications
Antigen HDAC1
Användningsområden ELISA, Immunohistochemistry (PFA fixed), Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of HDAC1.
Utspädning Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user.
Formulering In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide)
Gen HDAC1
Gene Alias DKFZp686H12203/GON-10/HD1/RPD3/RPD3L1
Gensymboler HDAC1
Värd art Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S421/S423 of human HDAC1.
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3065
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
For Research Use Only
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.