missing translation for 'onlineSavingsMsg'
Läs mer

HOXB9, Rabbit, Polyclonal Antibody, Abnova™

Produktkod. 16077337
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16077337 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16077337 Supplier Abnova Supplier No. PAB27154.100uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of HOXB9.

This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded nuclear protein functions as a sequence-specific transcription factor that is involved in cell proliferation and differentiation. Increased expression of this gene is associated with some cases of leukemia, prostate cancer and lung cancer. [provided by RefSeq

Specifications

Antigen HOXB9
Användningsområden Immunohistochemistry, Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of HOXB9.
Utspädning Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulering In PBS, pH 7.2 (0.05% sodium azide)
Gen HOXB9
Gene Alias HOX-2.5/HOX2/HOX2E
Gensymboler HOXB9
Värd art Rabbit
Immunogen A synthetic peptide corresponding to human HOXB9.
Reningsmetod Antigen affinity purification
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3219
Testspecificitet HOXB9 polyclonal antibody detects endogenous levels of HOXB9 protein.
Målarter Human, Mouse
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.