Läs mer
JAK2 (phospho Y221), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Beskrivning
This gene product is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways. It has been found to be constituitively associated with the prolactin receptor and is required for responses to gamma interferon. Mice that do not express an active protein for this gene exhibit embryonic lethality associated with the absence of definitive erythropoiesis. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | JAK2 |
| Användningsområden | Immunohistochemistry (PFA fixed) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic phosphopeptide of JAK2. |
| Utspädning | Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gen | JAK2 |
| Gene Alias | JTK10 |
| Gensymboler | JAK2 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.