Läs mer
JUN (phospho T239), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Descripción
This gene is the putative transforming gene of avian sarcoma virus 17. It encodes a protein which is highly similar to the viral protein, and which interacts directly with specific target DNA sequences to regulate gene expression. This gene is intronless and is mapped to 1p32-p31, a chromosomal region involved in both translocations and deletions in human malignancies. [provided by RefSeq
Especificaciones
Especificaciones
| Antigen | JUN |
| Användningsområden | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic phosphopeptide of JUN. |
| Utspädning | Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In TBS, pH 7.2 (BSA, 10% Proclin300) |
| Gen | JUN |
| Gene Alias | AP-1/AP1/c-Jun |
| Gensymboler | JUN |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.