missing translation for 'onlineSavingsMsg'
Läs mer

KCNMA1 Rabbit anti-Bovine, Chicken, Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Xenopus, Zebra fish, Polyclonal Antibody, Abnova™

Produktkod. 16047747
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μg
Förpackningsstorlek:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16047747 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16047747 Leverantör Abnova Leverantörsnummer PAB27755.50ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of KCNMA1.

MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit, which is the product of this gene, and the modulatory beta subunit. Intracellular calcium regulates the physical association between the alpha and beta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifikationer

Antigen KCNMA1
Användningsområden Immunohistochemistry (PFA fixed)
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of KCNMA1.
Utspädning Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.09% sodium azide)
Gen KCNMA1
Gene Alias BKTM/DKFZp686K1437/KCa1.1/MGC71881/MaxiK/SAKCA/SLO/SLO-ALPHA/SLO1/mSLO1
Gensymboler KCNMA1
Värd art Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acid at internal region of human KCNMA1.
Reningsmetod Immunoaffinity chromatography
Kvantitet 50 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3778
Testspecificitet BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Målarter Bovine, Canine, Chicken, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Xenopus species, Zebrafish
Innehåll och lagring Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.