Läs mer
KIAA1919, Rabbit, Polyclonal Antibody, Abnova™
Beskrivning
Sequence: SGLNEYEEENEEEDAEKWNEMDFEMIETNDTMRHSIIETSRSSLTEPTAEVYNQYPSNALVFESSPFNTGSAHVKHLPETR
Specifikationer
Specifikationer
| Antigen | KIAA1919 |
| Användningsområden | Immunohistochemistry (PFA fixed) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against recombinant KIAA1919. |
| Utspädning | Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) |
| Gen | KIAA1919 |
| Gene Alias | MGC33953/NaGLT1 |
| Gensymboler | KIAA1919 |
| Visa mer |
For Research Use Only