Läs mer
KIT Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KIT.
Brand: Abnova PAB3359.400uL
Beskrivning
This gene encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifikationer
| KIT | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KIT. | |
| In PBS (0.09% sodium azide) | |
| C-Kit/CD117/PBT/SCFR | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 3815 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Flow Cytometry (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| KIT | |
| KIT | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human KIT. | |
| 400 μL | |
| Primary | |
| Human, Monkey | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only