missing translation for 'onlineSavingsMsg'
Läs mer

LEFTY2, Rabbit, Clone: 7C5G1H6H10, Abnova™

Produktkod. 16095416
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16095416 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16095416 Leverantör Abnova Leverantörsnummer MAB2032.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit monoclonal antibody raised against recombinant LEFTY2.

This gene encodes a member of the TGF-beta family of proteins. The encoded protein is secreted and plays a role in left-right asymmetry determination of organ systems during development. The protein may also play a role in endometrial bleeding. Mutations in this gene have been associated with left-right axis malformations, particularly in the heart and lungs. Some types of infertility have been associated with dysregulated expression of this gene in the endometrium. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene. [provided by RefSeq

Specifikationer

Antigen LEFTY2
Användningsområden ELISA, Western Blot
Klassificering Monoclonal
Klona 7C5G1H6H10
Konjugera Unconjugated
Beskrivning Rabbit monoclonal antibody raised against recombinant LEFTY2.
Utspädning ELISA (1:5000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulering In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gen LEFTY2
Gene Alias EBAF/LEFTA/LEFTYA/MGC46222/TGFB4
Gensymboler LEFTY2
Värd art Rabbit
Immunogen Recombinant His fusion protein corresponding to human LEFTY2.
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 7044
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotyp IgG
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.