missing translation for 'onlineSavingsMsg'
Läs mer

MARK2 (phospho T595), Rabbit, Polyclonal Antibody, Abnova™

Produktkod. 16059196
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16059196 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16059196 Leverantör Abnova Leverantörsnummer PAB10332.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MARK2.

This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifikationer

Antigen MARK2
Användningsområden ELISA, Immunofluorescence, Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of MARK2.
Utspädning ELISA (1:1000-1:3000) Western Blot (1:250-1:1000) The optimal working dilution should be determined by the end user.
Formulering In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gen MARK2
Gene Alias EMK1/MGC99619/PAR-1/Par1b
Gensymboler MARK2
Värd art Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T595 of human MARK2.
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2011
Testspecificitet Reactivity occurs against human MARK2 pT595 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated human MARK2 is minimal by ELISA. Expect at least partial reactivity with other isoforms of MARK. The immunogen sequence is present in all forms identified to date and localizes to T595 on MARK2, T587 on MARK3, T591 on MARK1 and T568 on MARK4.
Målarter Bovine, Canine, Human, Mouse, Rat
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.