Läs mer
ME3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Beskrivning
Malic enzyme catalyzes the oxidative decarboxylation of malate to pyruvate using either NAD+ or NADP+ as a cofactor. Mammalian tissues contain 3 distinct isoforms of malic enzyme: a cytosolic NADP(+)-dependent isoform, a mitochondrial NADP(+)-dependent isoform, and a mitochondrial NAD(+)-dependent isoform. This gene encodes a mitochondrial NADP(+)-dependent isoform. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | ME3 |
| Användningsområden | Immunohistochemistry (PFA fixed), Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of ME3. |
| Utspädning | Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.2 (0.05% sodium azide) |
| Gen | ME3 |
| Gene Alias | FLJ34862 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.