missing translation for 'onlineSavingsMsg'
Läs mer

NCOA3, Rabbit, Polyclonal Antibody, Abnova™

Produktkod. 16099156
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16099156 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16099156 Leverantör Abnova Leverantörsnummer PAB10282.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of NCOA3.

The protein encoded by this gene is a nuclear receptor coactivator that interacts with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has histone acetyltransferase activity and recruits p300/CBP-associated factor and CREB binding protein as part of a multisubunit coactivation complex. This protein is initially found in the cytoplasm but is translocated into the nucleus upon phosphorylation. Two transcript variants encoding different isoforms have been found for this gene. In addition, a polymorphic repeat region is found in the C-terminus of the encoded protein. [provided by RefSeq

Specifikationer

Antigen NCOA3
Användningsområden ELISA, Immunoprecipitation, Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of NCOA3.
Utspädning ELISA (1:175000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulering In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gen NCOA3
Gene Alias ACTR/AIB-1/AIB1/CAGH16/CTG26/KAT13B/MGC141848/RAC3/SRC3/TNRC14/TNRC16/TRAM-1/bHLHe42/pCIP
Gensymboler NCOA3
Värd art Rabbit
Immunogen A synthetic peptide corresponding to internal region of human NCOA3.
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 8202
Testspecificitet This antibody reacts with several isoforms of human NCOA3 protein (a, b, d, e and f).
Målarter Bovine, Canine, Human, Macaque, Mouse, Rat
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.