Läs mer
NTN1, Rabbit, Polyclonal Antibody, Abnova™
Beskrivning
Netrin is included in a family of laminin-related secreted proteins. The function of this gene has not yet been defined; however, netrin is thought to be involved in axon guidance and cell migration during development. Mutations and loss of expression of netrin suggest that variation in netrin may be involved in cancer development. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | NTN1 |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of NTN1. |
| Utspädning | Western blot (0.5 to 2 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.08% sodium azide) |
| Gen | NTN1 |
| Gene Alias | NTN1L |
| Gensymboler | NTN1 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.