Läs mer
P2RX3, Rabbit, Polyclonal Antibody, Abnova™
Beskrivning
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and may transduce ATP-evoked nociceptor activation. Mouse studies suggest that this receptor is important for peripheral pain responses, and also participates in pathways controlling urinary bladder volume reflexes. It is possible that the development of selective antagonists for this receptor may have a therapeutic potential in pain relief and in the treatment of disorders of urine storage. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | P2RX3 |
| Användningsområden | Immunohistochemistry (PFA fixed) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of P2RX3. |
| Utspädning | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gen | P2RX3 |
| Gene Alias | MGC129956/P2X3 |
| Gensymboler | P2RX3 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.