missing translation for 'onlineSavingsMsg'
Läs mer

PAK6 (phospho S165), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16080718
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16080718 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16080718 Leverantör Abnova Leverantörsnummer PAB7902.100uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK6.

This gene encodes a member of the p21-activated kinase (PAK) family. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR), which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development. This gene was found to be highly expressed in testis and prostate tissues and the encoded protein was shown to cotranslocate into the nucleus with AR in response to androgen. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifikationer

Antigen PAK6
Användningsområden ELISA, Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK6.
Utspädning ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulering In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gen PAK6
Gene Alias PAK5
Gensymboler PAK6
Värd art Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S165 of human PAK6.
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 56924
Testspecificitet The sequence used has two amino acid differences compared to rat and mouse PAK6.
Målarter Human
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.