Learn More
PCDH18, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PCDH18.
Brand: Abnova PAB16718.100ug
Additional Details : Weight : 0.02000kg
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. This gene encodes a protein which contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Although its specific function is undetermined, the cadherin-related neuronal receptor is thought to play a role in the establishment and function of specific cell-cell connections in the brain. [provided by RefSeq
Specifications
PCDH18 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PCDH18. | |
In PBS (0.02% sodium azide) | |
DKFZp434B0923/KIAA1562/PCDH68L | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
PCDH18 | |
PCDH18 | |
A synthetic peptide corresponding to N-terminus 17 amino acids of human PCDH18. | |
RUO | |
54510 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only