Läs mer
PTGES Rabbit anti-Bovine, Chicken, Dog, Horse, Human, Monkey, Pig, Rabbit, Rat, Zebra fish, Polyclonal Antibody, Abnova™
Beskrivning
The protein encoded by this gene is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis. Knockout studies in mice suggest that this gene may contribute to the pathogenesis of collagen-induced arthritis and mediate acute pain during inflammatory responses. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | PTGES |
| Användningsområden | Immunohistochemistry (PFA fixed) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of PTGES. |
| Utspädning | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.09% sodium azide) |
| Gen | PTGES |
| Gene Alias | MGC10317/MGST-IV/MGST1-L1/MGST1L1/MPGES/PGES/PIG12/PP102/PP1294/TP53I12/mPGES-1 |
| Gensymboler | PTGES |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.