missing translation for 'onlineSavingsMsg'
Läs mer

RAF1 (phospho T269), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16058856
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
400 μL
Förpackningsstorlek:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16058856 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16058856 Leverantör Abnova Leverantörsnummer PAB0594.400uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAF1.

This gene is the cellular homolog of viral raf gene (v-raf). The encoded protein is a MAP kinase kinase kinase (MAP3K), which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated, the cellular RAF1 protein can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2, which in turn phosphorylate to activate the serine/threonine specific protein kinases, ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2. [provided by RefSeq

Specifikationer

Antigen RAF1
Användningsområden Dot Blot, ELISA
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAF1.
Utspädning Dot Blot (1:100-500) The optimal working dilution should be determined by the end user.
Formulering In PBS (0.09% sodium azide)
Gen RAF1
Gene Alias CRAF/NS5/Raf-1/c-Raf
Gensymboler RAF1
Värd art Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T269 of human RAF1.
Reningsmetod Protein A purification
Kvantitet 400 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 5894
Målarter Human
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.