Läs mer
RPS6KB1 (phospho S434), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Beskrivning
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding this gene and overexpression of this kinase are seen in some breast cancer cell lines. Alternate translational start sites have been described and alternate transcriptional splice variants have been observed but have not been thoroughly characterized. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | RPS6KB1 |
| Användningsområden | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic phosphopeptide of RPS6KB1. |
| Utspädning | Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In TBS, pH 7.2 (BSA, 10% Proclin300) |
| Gen | RPS6KB1 |
| Gene Alias | PS6K/S6K/S6K1/STK14A/p70(S6K)-alpha/p70-S6K/p70-alpha |
| Gensymboler | RPS6KB1 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.