Läs mer
RSHL3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Beskrivning
This gene encodes a protein that appears to be a component the radial spoke head, as determined by homology to similar proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates. Radial spokes, which are regularly spaced along cilia, sperm, and flagella axonemes, consist of a thin 'stalk' and a bulbous 'head' that form a signal transduction scaffold between the central pair of microtubules and dynein. Mutations in this gene cause primary ciliary dyskinesia 1, a disease arising from dysmotility of motile cilia and sperm. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | RSHL3 |
| Användningsområden | Immunofluorescence, Immunohistochemistry, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against recombinant RSHL3. |
| Utspädning | Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) |
| Gen | RSHL3 |
| Gene Alias | FLJ37974/MGC126303/dJ412I7.1 |
| Gensymboler | RSHL3 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.