Learn More
TAF1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TAF1.
Brand: Abnova PAB11285.100ug
Additional Details : Weight : 0.02000kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is the basal transcription factor TFIID, which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes the largest subunit of TFIID. This subunit binds to core promoter sequences encompassing the transcription start site. It also binds to activators and other transcriptional regulators, and these interactions affect the rate of transcription initiation. This subunit contains two independent protein kinase domains at the N and C-terminals, but also possesses acetyltransferase activity and can act as a ubiquitin-activating/conjugating enzyme. This gene is part of a complex transcriptional unit (TAF1/DYT3), wherein some products share exons with TAF1 as well as additional exons downstream.[provided by RefSeq
Specifications
TAF1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TAF1. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
BA2R/CCG1/CCGS/DYT3/KAT4/N-TAF1/NSCL2/OF/P250/TAF2A/TAFII250 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:10000-1:50000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. | |
TAF1 | |
TAF1 | |
A synthetic peptide corresponding to C-terminus of human TAF1. | |
RUO | |
6872 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only