Läs mer
TANK Rabbit anti-Human, Polyclonal Antibody, Abnova™
Beskrivning
The TRAF (tumor necrosis factor receptor-associated factor) family of proteins associate with and transduce signals from members of the tumor necrosis factor receptor superfamily. The protein encoded by this gene is found in the cytoplasm and can bind to TRAF1, TRAF2, or TRAF3, thereby inhibiting TRAF function by sequestering the TRAFs in a latent state in the cytoplasm. For example, the protein encoded by this gene can block TRAF2 binding to LMP1, the Epstein-Barr virus transforming protein, and inhibit LMP1-mediated NF-kappa-B activation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | TANK |
| Användningsområden | Immunocytochemistry, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of TANK. |
| Utspädning | Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.02% sodium azide) |
| Gen | TANK |
| Gene Alias | I-TRAF/TRAF2 |
| Gensymboler | TANK |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.